Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 1,2,3-benzothiadiazole-5-carboxylate
Methyl 1,2,3-benzothiadiazole-5-carboxylate
Methyl 1,2,3-benzothiadiazole-5-carboxylate
Irritant
Methyl 1,2,3-benzothiadiazole-5-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 1,2,3-benzothiadiazole-5-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
23616-15-1
:
Formula:
C₈H₆N₂O₂S
MDL Number:
MFCD01566550
Melting Pt:
118…120 °C
:
194.21 g/mol
:
GHS07
Storage Temperature:
Ambient