Ethyl 5-(4-chlorophenyl)-4-(chlorosulphonyl)-2-methyl-3-furoate, tech
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306936-30-1
:
Formula:C₁₄H₁₂Cl₂O₅S
MDL Number:MFCD01934803
:363.213 g/mol
Melting Pt:79…81 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05