Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
t-Boc-N-Amido-PEG6-Amine
t-Boc-N-Amido-PEG6-Amine
t-Boc-N-Amido-PEG6-Amine
t-Boc-N-Amido-PEG6-Amine
Supplier:
Apollo Scientific
Certificates
Expand image
t-Boc-N-Amido-PEG6-Amine
Supplier:
Apollo Scientific
About this item
CAS Number:
1091627-77-8
:
Formula:
C₁₉H₄₀N₂O₈
MDL Number:
MFCD16619391
:
424.529 g/mol
:
GHS07
Storage Temperature:
Refrigerator