Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
t-Boc-N-Amido-PEG5-Amine
t-Boc-N-Amido-PEG5-Amine
t-Boc-N-Amido-PEG5-Amine
Irritant
t-Boc-N-Amido-PEG5-Amine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
t-Boc-N-Amido-PEG5-Amine
Supplier:
Apollo Scientific
About this item
CAS Number:
189209-27-6
:
Formula:
C₁₇H₃₆N₂O₇
MDL Number:
MFCD22056313
:
380.477 g/mol
:
GHS07
Storage Temperature:
Refrigerator