Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin-PEG4-PFP ester
Biotin-PEG4-PFP ester
Biotin-PEG4-PFP ester
APOSBIPG1233-100MG
:
Apollo Scientific
:
Biotin-PEG4-PFP ester
APOSBIPG1233-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
657.647 g/mol
:
GHS07
:
Refrigerator
:
C₂₇H₃₆F₅N₃O₈S
:
MFCD13184979