Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
rac-GR24
rac-GR24
rac-GR24
Irritant
rac-GR24
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
rac-GR24
Supplier:
Apollo Scientific
About this item
CAS Number:
76974-79-3
:
Formula:
C₁₇H₁₄O₅
MDL Number:
MFCD12405021
:
298.29 g/mol
Melting Pt:
156…157 °C
Density:
1.364 g/cm³ (25 °C)
:
GHS07
Storage Temperature:
Freezer