Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-O-Benzylidene-D-arabinitol
1,3-O-Benzylidene-D-arabinitol ≥98%
1,3-O-Benzylidene-D-arabinitol ≥98%
1,3-O-Benzylidene-D-arabinitol ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-O-Benzylidene-D-arabinitol ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
70831-50-4
:
Formula:
C₁₂H₁₆O₅
MDL Number:
MFCD00049363
:
240.252 g/mol
Melting Pt:
128…130 °C
:
GHS07
Storage Temperature:
Freezer