NM522 competent cells
NM522 competent cells, 5×0,2 ml; 1×10⁸ transformants/µg, 5×0,2 ml
AGLS200233
: AGILENT
NM522 competent cells are recombination-proficient and EcoK restriction-deficient.
- Recombination-proficient and EcoK restriction-deficient
- Blue-white colour screening possible for recombinant plasmids
- Transformation efficiency >1×10⁸ transformants/µg DNA
NM6522 competent cells are a classic E. coli strain for economical cloning when you don’t need supercompetent efficiencies. NM522 is a recombination-proficient and EcoK restriction deficient host strain. Intrinsic antibiotic resistance aids cell selection after a complete and successful transformation.
: For research use only. Not for use in diagnostic procedures.
- Description:NM522 competent cells
- Seq:01
- Pk Info:5×0,2 ml
- Feature:5×0,2 ml; 1×10⁸ transformants/µg