EPZ-6438 95%
Catalog # ACRO468171000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to off-white Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:572.74 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1403254-99-8
:MFCD24849415
Formula:C₃₄H₄₄N₄O₄
MDL Number:MFCD20527867