Orders LinkContactUsLinkComponent
ABLE C competent cells
ABLE C competent cells
Marketsource
Supplier:   STRATAGENE
ABLE C competent cells
Supplier:  STRATAGENE
Marketsource
Certificates
ABLE C competent cells
6 Options Available Below
Product Details & Documents
ABLE Competent Cells for toxic clones reduce copy number four to ten fold compared to the XL1-Blue strain.
  • Reduces the copy number of ColE1-derived plasmids (such as pUC and pBluescript® plasmids) four-fold
  • Decreases level of cloned protein product to increase cell viability
  • Transformation efficiency of 5×10⁶ to 2×10⁷ transformants/µg
  • Also available in an electroporation-competent version
ABLE C Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE C strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
The ABLE C Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE C strains.
ABLE K Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE K strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
ABLE K Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE K strains.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Feature
    Pk Info
    Availability
    Price
  • STRA200161
    ABLE C electroporation-competent cells
    ABLE C electroporation-competent cells
    Reduces the number of common cloning vectors 4-fold
    5×0,1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 570,00 €
    570,00 €
    1 * 1 ST
       570,00 €
    Specifications:
    Marketsource
    Marketsource
  • STRA200162
    ABLE K electroporation-competent cells
    ABLE K electroporation-competent cells
    Reduces the number of common cloning vectors 10-fold.
    5×0,1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 570,00 €
    570,00 €
    1 * 1 ST
       570,00 €
    Specifications:
    Marketsource
    Marketsource
  • STRA200171
    ABLE C competent cells
    ABLE C competent cells
    Reduces the number of common cloning vectors 4-fold
    5×0,2 ml
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 570,00 €
    570,00 €
    1 * 1 mL
       570,00 €
    Specifications:
    Marketsource
    Marketsource
  • STRA200170
    ABLE competent cell kit
    ABLE competent cell kit
    Includes both ABLE C and K cells, Contains: 5×0.2 ml ABLE C and 5×0.2 ml ABLE K cells.
    1 kit
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 923,00 €
    923,00 €
    1 * 1 KIT
       923,00 €
    Specifications:
    Marketsource
    Marketsource
  • STRA200160
    ABLE electroporation-competent cell kit
    ABLE electroporation-competent cell kit
    Includes both ABLE C and K cells, Contains: 5×0.1 ml ABLE C cells, 5×0.1 ml ABLE K cells.
    1 kit
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 923,00 €
    923,00 €
    1 * 1 KIT
       923,00 €
    Specifications:
    Marketsource
    Marketsource
  • STRA200172
    ABLE K competent cells
    ABLE K competent cells
    Reduces copy number of common cloning vectors by 10-fold, allowing toxic clones to replicate.
    5×0,2 ml
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 570,00 €
    570,00 €
    1 * 1 mL
       570,00 €
    Specifications:
    Marketsource
    Marketsource