Trichloroacetic acid (TCA) 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Boiling Pt:196 °C
Melting Pt:52…58 °C
Density:1.62g/cm³ (25 °C)
Flash Pt:>113 °C
Storage Temperature:Ambient
EINECS:200-927-2
UN:1839
ADR:8,II
:GHS05
:GHS09