Order Entry
Germany
Orders LinkContactUsLinkComponent
Affinity chromatography columns, StrepTrap™ XT

Affinity chromatography columns, StrepTrap™ XT

Supplier:   Cytiva
Affinity chromatography columns, StrepTrap™ XT
Supplier:  Cytiva
Certificates
Affinity chromatography columns, StrepTrap™ XT
Affinity chromatography columns, StrepTrap™ XT

4 Options Available Below

Product Details & Documents

A chromatography resin for the purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein.

  • Highly pure Strep-tag® II and Twin-Strep-tag® recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration

Compatible with a wide range of detergents, denaturants, and other additives. Available in 10 and 50 ml lab packs of bulk resin and prepacked in 1 and 5 ml StrepTrap™ XT chromatography columns for convenience.

Specifications for Product Family

Specifications

  • Catalog No:
  • 29-4013-17
  • 29-4013-20
  • 29-4013-22
  • 29-4013-23
  • Application:
  • Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
  • Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
  • Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
  • Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 6 - 10
  • 6 - 10
  • 6 - 10
  • 6 - 10
  • Storage:
  • 2...8 °C in 20% ethanol
  • 2...8 °C in 20% ethanol
  • 2...8 °C in 20% ethanol
  • 2...8 °C in 20% ethanol

Specifications

Product Family Options

Product Information

  • Column hardware
    Particle Size
    Ø int.
    Length
    Capacity
    Flow rate
    Availability
    Price
  • 29-4013-17
    StrepTrap™ XT
    StrepTrap™ XT
    PP
    34 µm
    7,00 mm
    25 mm
    1×1 ml
    1 ml/min
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 245,00 €
    245,00 €
    1 * 1 mL
       245,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    29-4013-17
    Application
    Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
    Max. pressure
    5 bar
    pH range
    6 - 10
    Storage
    2...8 °C in 20% ethanol
  • 29-4013-20
    StrepTrap™ XT
    StrepTrap™ XT
    PP
    34 µm
    7,00 mm
    25 mm
    5×1 ml
    1 ml/min
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 615,00 €
    615,00 €
    1 * 5 ST
       615,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    29-4013-20
    Application
    Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
    Max. pressure
    5 bar
    pH range
    6 - 10
    Storage
    2...8 °C in 20% ethanol
  • 29-4013-22
    StrepTrap™ XT
    StrepTrap™ XT
    PP
    34 µm
    16,00 mm
    25 mm
    1×5 ml
    5 ml/min
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 604,00 €
    604,00 €
    1 * 5 mL
       604,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    29-4013-22
    Application
    Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
    Max. pressure
    5 bar
    pH range
    6 - 10
    Storage
    2...8 °C in 20% ethanol
  • 29-4013-23
    StrepTrap™ XT
    StrepTrap™ XT
    PP
    34 µm
    16,00 mm
    25 mm
    5×5 ml
    5 ml/min
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 2 540,00 €
    2 540,00 €
    1 * 5 ST
       2 540,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    29-4013-23
    Application
    Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
    Max. pressure
    5 bar
    pH range
    6 - 10
    Storage
    2...8 °C in 20% ethanol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.