Nitric acid 1 mol/l (1 N), Titripur®, Supelco®
Catalog # 1.60307.1003
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle with RFID tag
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:63.01 g/mol
ADR:8,
:M-Clarity™ quality level = MQ100
UN:2031
:GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349