Order Entry
Germany
ContactUsLinkComponent
Methylene blue (Neisser) for microscopy, Sigma-Aldrich®
Methylene blue (Neisser) for microscopy, Sigma-Aldrich®
Catalog # 01363-100ML-F
CAS Number:  
Methylene blue (Neisser) for microscopy, Sigma-Aldrich®
Catalog # 01363-100ML-F
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :319.86 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:61-73-4
 :MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008