1,1′-Dioctadecyl-4,4′-bipyridinium dibromide ≥97%, Sigma-Aldrich®
Catalog # 391506-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:210…220 °C (dec., lit.)
:822.96 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:90179-58-1
Formula:C₄₆H₈₂Br₂N₂
MDL Number:MFCD00192099