1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt, Sigma-Aldrich®
456063-25G
:
- Pk:25 g
- Seq:3308
- Appearance (Colour):Orange to Red
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Titration with AgNO₃ After Oxygen Combustion:Specification: Report Results
- Purity (HPLC):≥83.0 %
:
:404.17 g/mol
:>300 °C (lit.)
:M-Clarity™ quality level = MQ100
:228-391-5
:GHS07
:Room Temperature
:6258-06-6
:C₁₄H₈BrNO₅S
:MFCD00019160