Xylylazo violet I, Sigma-Aldrich®
Catalog # 248266-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:513.50 g/mol
Storage Temperature:Ambient
CAS Number:14936-97-1
Formula:C₂₅H₂₁N₃O₆S
MDL Number:MFCD00003890