Order Entry
Germany
ContactUsLinkComponent
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3), Sigma-Aldrich®
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3), Sigma-Aldrich®
Catalog # 309052-25G
CAS Number:  
undefined
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3), Sigma-Aldrich®
Catalog # 309052-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:8004-92-0
Formula:C₁₈H₁₁NO₂.xNaO₃S
MDL Number:MFCD00080727