Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acid blue 25
Acid blue 25 45% (dye content), Sigma-Aldrich®
Acid blue 25 45% (dye content), Sigma-Aldrich®
210684-25G
:
SIGMA ALDRICH MICROSCOPY
:
Acid blue 25 45% (dye content), Sigma-Aldrich®
210684-25G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
25 g
Seq:
3308
:
:
416.4 g/mol
:
GHS07
:
Ambient
:
6408-78-2
:
C₂₀H₁₃N₂O₅SNa
:
MFCD00001214