Order Entry
Germany
ContactUsLinkComponent
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Sigma-Aldrich®
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Sigma-Aldrich®
Certificates
undefined
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    White to Light Brown
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Colorless to Light Yellow
  • Solubility (Turbidity)
    2.5 mg/mL, H2O:
    Clear
  • Proton NMR spectra:
    Conforms to Structure
  • Carbon:
    39.0 - 49.0 %
  • Nitrogen:
    5.5 - 7.5 %

Specification Test Results