Order Entry
Germany
ContactUsLinkComponent
Crystal violet solution, Sigma-Aldrich®
Flammable
Health
Irritant
Environment
Crystal violet solution, Sigma-Aldrich®
Catalog # HT90132-1L
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Crystal violet solution, Sigma-Aldrich®
Catalog # HT90132-1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :407.99 g/mol
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
 :1
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750