Order Entry
Germany
ContactUsLinkComponent
SynaptoRed™ C2 ≥95% (by HPLC), Sigma-Aldrich®
SynaptoRed™ C2 ≥95% (by HPLC), Sigma-Aldrich®
Catalog # S6689-5MG
CAS Number:  
undefined
SynaptoRed™ C2 ≥95% (by HPLC), Sigma-Aldrich®
Catalog # S6689-5MG
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :607.51 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:162112-35-8
Formula:C₃₀H₄₅Br₂N₃
MDL Number:MFCD00467819