Order Entry
Germany
ContactUsLinkComponent
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95% (by HPLC), Sigma-Aldrich®
Irritant
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95% (by HPLC), Sigma-Aldrich®
Catalog # D0940-500MG
CAS Number:  
undefined
Irritant
p-Xylene-bis(N-pyridinium bromide) (DPX) ≥95% (by HPLC), Sigma-Aldrich®
Catalog # D0940-500MG
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :422.16 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:14208-10-7
Formula:C₁₈H₁₈Br₂N₂
MDL Number:MFCD00043749