Biotin-4-fluorescein, Sigma-Aldrich®
Catalog # B9431-5MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:644.7 g/mol
Storage Temperature:Ambient
CAS Number:1032732-74-3
Formula:C₃₃H₃₂N₄O₈S