Order Entry
Germany
ContactUsLinkComponent
 
 
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, fluorogenic, Sigma-Aldrich®
 
undefined
 
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, fluorogenic, Sigma-Aldrich®

 

  • Appearance (Colour):
    White to Orange to Red
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    36.4 - 38.3%
  • Nitrogen:
    12.0 - 12.8%
  • Purity (HPLC):
    ≥98%

 

Frequently Bought Together