Order Entry
Germany
ContactUsLinkComponent
Dihydroethidium ≥95%, Sigma-Aldrich®
Dihydroethidium ≥95%, Sigma-Aldrich®
Catalog # D7008-10MG
CAS Number:  
undefined
Dihydroethidium ≥95%, Sigma-Aldrich®
Catalog # D7008-10MG
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :315.412 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:104821-25-2
 :38483-26-0
Formula:C₂₁H₂₁N₃
MDL Number:MFCD00077335