Order Entry
Germany
ContactUsLinkComponent
Pig Pepsin (from Stomach)
Pig Pepsin (from Stomach)
Certificates
Pig Pepsin (from Stomach)

Specifications

  • Source:
    Stomach
  • Species:
    Pig
  • Storage conditions:
    2…8 °C
  • Enzyme name:
    Pepsin
  • M-Clarity quality level:
    MQ100
  • Concentration:
    Ready to Use

Specifications