Order Entry
Germany
ContactUsLinkComponent
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-BI and fast red violet LB, Sigma-Aldrich®
Corrosive
Irritant
Health
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-BI and fast red violet LB, Sigma-Aldrich®
Catalog # 86R-1KT
CAS Number:  
Corrosive
Irritant
Health
Leukocyte Alkaline Phosphatase Kit based on naphthol AS-BI and fast red violet LB, Sigma-Aldrich®
Catalog # 86R-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
UN:3316
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
 :1