
Dexmedetomidine hydrochloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:145108-58-3
:
Formula:C₁₃H₁₆N₂·HCl
MDL Number:MFCD12912677
:236.74 g/mol
Melting Pt:158 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Melting point:156 to 160 °C
- HPLC:≥97.5 %
- Specific optical rotation:+48° to +54° (20 °C, 589 nm) (1% in water)