Order Entry
Germany
ContactUsLinkComponent
 
Cyproheptadine hydrochloride sesquihydrate
  ACRO462670050
 :  
undefined
Cyproheptadine hydrochloride sesquihydrate
  ACRO462670050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    7.0 to 9.0 %
  • Solubility:
    (5 % in ethanol) clear colorless to light yellow
  • Titration with HClO4:
    98.5 to 100.5 %

 

 

 :
 :350.88 g/mol
 :165 °C
 :Ambient
 :2811
 :6.1,III
 :GHS06
 :41354-29-4
 :C₂₁H₂₁N·HCl·11/2H₂O
 :MFCD00012538