N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:183319-69-9
:
Formula:C₂₂H₂₄ClN₃O₄
MDL Number:MFCD07781272
:429.901 g/mol
Melting Pt:215…227 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Solubility:(1% in DMSO) Clear solution
- HPLC:≥97.5 %
- Residue after ignition:≤0.5 %