Order Entry
Germany
ContactUsLinkComponent
Potassium dihydrogen phosphate for HPLC
Potassium dihydrogen phosphate for HPLC
Certificates
undefined
Potassium dihydrogen phosphate for HPLC

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Solubility:
    (0.1 g/ml) Clear colorless
  • UV:
    (0.5 M in water)
  • Titration with NaOH:
    ≥99.4 %
  • Gradient analysis:
    passes test

Specification Test Results