Order Entry
Germany
ContactUsLinkComponent
Ridascreen® Giardia
Ridascreen® Giardia
Catalog # RBIOC1101
Supplier:  R BIOPHARM
CAS Number:  
Ridascreen® Giardia
Catalog # RBIOC1101
Supplier:  R BIOPHARM
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Giardia Elisa tests (96)

Specifications

About this item

The RIDASCREEN® Giardia test is an enzyme immunoassay for the qualitative determination of Giardia lamblia in stool sample.

Giardia lamblia are flagellate parasites that cause infections in the small intestines of humans after the oral uptake of Giardia cysts. Because it is found worldwide, the Giardia lamblia pathogen is a significant cause of chronic diarrhea, in particular in medical cases related to travel. The infection occurs after ingestion of the cysts in contaminated food or water.

Specifications
Plate format: Microtiter plate with 96 wells (12 strips with 8 removable wells each)
Sensitivity: 85 % (compared to microscopy) - 96 % (compared to PCR)
Specificity: 93 % (compared to microscopy) - 100 % (compared to PCR)
Test time: 60 min (incubation)

Accessory: RIDASCREEN® Giardia Reference Controls (Art. No.: CRP1104)