Order Entry
Germany
ContactUsLinkComponent
n-Nonane ≥95.0%, PESTINORM®
Flammable
Health
Environment
n-Nonane ≥95.0%, PESTINORM®
Catalog # 85931.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Environment
n-Nonane ≥95.0%, PESTINORM®
Catalog # 85931.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    min. 95.00%
  • Evaporation residue :
    max. 1 ppm
  • Water :
    max. 0.0050 %
  • Pesticide analysis (Lindane/ECD):
    max. 5 ng/l

Specification Test Results

About this item

 :
ADR:3,III
 :Packaged under nitrogen.
Flash Pt:31 °C
 :128.26 g/mol
EINECS:203-913-4
UN:1920
 :GHS02
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:111-84-2
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574