Order Entry
Germany
ContactUsLinkComponent
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Catalog # 391-0637
Supplier:  VWR Collection
undefined
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Catalog # 391-0637
Supplier:  VWR Collection

Specifications

  • Optical property:
    Light blocking
  • Sterile:
    -
  • Description:
    1×8 sealing strips
  • Thickness:
    87,6 µm
  • Pk:
    300

Specifications

About this item

Easier to apply and remove as compared to standard moulded PCR strip caps.

  • Ideal for PCR
  • User-friendly alternative to strip-caps; no sore thumbs and fingers guaranteed
  • Seals a single 8-well row of a 96-well plate or a single PCR 8-tube strip
  • Certified DNase- and RNase-free
  • 2,0 mil pierceable foil
  • Recommended temperature range from −20 to +120 °C

6 strips are pre-cut on a continuous liner for ease of use.