Order Entry
Germany
ContactUsLinkComponent
VWR®, Reagent Reservoir, 175 ml
VWR®, Reagent Reservoir, 175 ml
Catalog # 391-0625
Supplier:  VWR Collection
undefined
VWR®, Reagent Reservoir, 175 ml
Catalog # 391-0625
Supplier:  VWR Collection

Specifications

  • Colour:
    Transparent
  • Description:
    Reagent reservoir
  • Pk:
    10

Specifications

About this item

175 ml graduated polypropylene reagent reservoir with lid. Unique V-shaped bottom reduces dead volume.

  • Easy open locking lid
  • Graduation marks at 25, 50, 75, 100, 125 and 150 ml
  • Excellent chemical resistance (PP)
  • Stackable

Can be autoclaved.