

1-Methylimidazole (NMI), anhydrous, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:616-47-7
:
Formula:C₄H₆N₂
MDL Number:MFCD00005292
Boiling Pt:198 °C
Melting Pt:–2 °C
Density:1.03 g/cm³
Flash Pt:92 °C
UN:2922
ADR:8,II
:82.11 g/mol
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear colorless Liquid (May discolor to yellow on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4955 to 1.4975 (20°C, 589 nm)
- Water:≤0.005 %
- GC:≥98.5 %