Restriction enzymes, Eco32I (EcoRV) (HC), Fermentas
Catalog # ER0303
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:50 U/µl
- Pk:1 KIT
- Compatible buffer:FastDigest Green Buffer or FastDigest Buffer (colorless)
- Overhang type:Blunt
- Description:Eco32I (EcoRV) (HC)
- Enzyme name:EcoRV
- Units:1000 units
Specifications
About this item
Eco32I (EcoRV) restriction enzyme recognizes GAT^ATC sites and cuts best at 37 °C in R buffer Isoschizomers: EcoRV.
- Superior quality—stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Used in molecular cloning, restriction site mapping, genotyping, southern blotting and restriction fragment length polymorphism (RFLP).