Hand disinfectants, Sterillium® Pure
Supplier: Bode
Certificates
About this item
The first ECARF-certified hand sanitizer. Without dyes and perfumes and with a proven skin care complex.
- Broad efficacy spectrum, including limited virucidal PLUS
- Inactivates noroviruses within hygienic hand disinfection
- ECARF-certified: particularly suitable for sensitive skin, asthmatics and atopics
- Increases skin moisture with regular use
- No impairment of the suitability for consumption of food (HACCP evaluated)
Required spectrum of activity SARS-CoV-2: limited virucidal
All disinfectants from HARTMANN for hands, skin, surfaces and instruments meet the effectiveness requirements for limited virucidal properties and can be used.
* tested on murine norovirus