Desiccator cabinet, SICCO Big Star
Catalog # 467-0061
Supplier: Bohlender
Specifications
- W×D×H ext.:560×580×560 mm
- Description:Big Star for drying and storage (156 L)
- Pk:1
- Shelves:2
- W×D×H int.:495×540×500 mm
Specifications
About this item
This transparent acrylic cabinet has a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed for storing or drying humidity sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA samples.
- Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
- Temperature resistance from −20 to +70 °C
- Cabinet has non slip rubber feet and is stackable
Delivery: Supplied with two removable, stainless steel, telescopic shelves and desiccant tray. Silica gel not included. Capacity to accommodate a maximum of 8 shelves.