Blotting membranes, Amersham™ Hybond™-N+
Supplier: Cytiva
Certificates
About this item
Amersham™ Hybond™-N+ is a positively charged nylon membrane with a binding capacity for nucleic acids up to 600 µg/cm² recommended for use with radioactive or non radioactive chemiluminescence and chemifluorescence detection systems.
- Recommended for all high density membrane arrays
- Multiple reprobing is possible without loss of membrane integrity
- QC tested in genomic Southern blots
Amersham™ Hybond™-N+ yields excellent sensitivity in conventional Southern blotting. Nucleic acid samples may be fixed by UV exposure or baking, although UV fixation is best for maximum reproducibility.
Pore size: 0,45 µm
Nucleic acid size required: >50 bp
*designed to fit Nunc™ OmniTray.
Caution: Amersham™ Hybond™-N+ is not suitable for alkali blotting and fixation.
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements