VWR® Reagent Bottles, Wide Neck
Supplier: VWR Collection
Certificates
About this item
Made from clear or amber soda-lime glass. Ideal for sampling, storage of powders or liquids, or other applications that do not involve high thermal stress.
Good chemical and physical properties
- Clear or amber glass, available with or without black PP screw cap
- Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Certifications: Thread in accordance with DIN 168.