ContactUsLinkComponent
Water bath disinfection reagent, Aquabator-Clean™
Water bath disinfection reagent, Aquabator-Clean™
Catalog # A9390.0250
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Water bath disinfection reagent, Aquabator-Clean™
Catalog # A9390.0250
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Pk:
    250 mL
  • Description:
    Aquabator-Clean™ (100X)
  • For:
    Disinfection of water baths
Specifications
About this item
Aquabator-Clean™ is designed for disinfecting water baths. It protects against growth of fungi and bacteria.
  • Supplied as a concentrate - 10 ml per litre of water
  • Biodegradable
  • Non irrtating to skin at recommended concentrations
Caution: Use biocides safely. Always read the label and product information before use.
Not suitable for CO₂ incubators.