ContactUsLinkComponent
Screw gate carabiner, Steel, Protecta®, 3M™
Screw gate carabiner, Steel, Protecta®, 3M™
Catalog # CPSDAJ501
Supplier:  CAPITAL SAFETY
undefined
Screw gate carabiner, Steel, Protecta®, 3M™
Catalog # CPSDAJ501
Supplier:  CAPITAL SAFETY
Specifications
  • Description:
    Screw gate carabiner, steel
  • Pk:
    20
Specifications
About this item
The Protecta® Screw gate carabiner is ideal in working situations where opening is occasional during the working day, as the gate can be opened by pushing it onto the anchorage point. The gate can then be manually locked once the carabiner holds the weight of the device.
  • Manual locking design is ideal for occasional opening during the working week
  • For use in all working at height applications
  • Durable and lightweight
  • Corrosion-resistant
Anchorage connectors are a critical component in a fall protection system, as they securely attach the user's system to the anchorage. The choice of anchor depends on the type of work being done. Anchorages are selected by what they connect to (general/all purpose, steel, concrete, roof, or specialty), whether they are fixed or mobile, and whether they are permanent or portable.
This device is suitable for use in mining, general industrial, construction, oil and gas, and transportation industries.
Weight: 0,17 kg.
Certifications: EN 362