Maintenance socks, PW® Spill, SM10
Catalog # PRTWSM10GRR
Supplier: Portwest
Specifications
- Description:Spill maintenance socks, grey
- Pk:40
Specifications
About this item
The maintenance socks have a knitted outer skin with fast absorbency speeds. It absorbs more than 4,5 litres of water and oil-based liquids.
- Made of polyester knitted fabric outer with cellulous fill
These products are grey in colour and will absorb all kinds of industrial liquids including oil based and water based spills. Ideal for use in industrial situations where many different liquids are present at the same time.
Certifications: BS 7959-1; BS 7959-3.