Welding safety spectacle, PW68
Catalog # PRTWPW68BGR
Supplier: Portwest
Specifications
- Colour:Shade 5
- Description:Welding safety spectacle, bottle green
- Pk:1
Specifications
About this item
Multipurpose Shade 5 lens with lateral protection and adjustable arms for customised comfort.
- 99% UV protection
- Anti-scratch coating for added durability
- Free spec cord
- Adjustable length for a secure and comfortable fit
- Excellent side protection
- Ultra lightweight for long uses
- Individually packed for vending machines
- Retail bag which aids presentation for retail sales