Chemical resistant gloves, Nitrosafe A810
Supplier: Portwest
Certificates
About this item
Chemical resistant gauntlet with textured pattern for enhanced grip.
- Textured pattern for enhanced grip
- Fully coated for maximum liquid protection
- Flock lined for added comfort
For use in the chemical, oil and food industries.
Certifications: EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X); AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .10.1
Specifications
- Catalog No:
- PRTWA810GNRXL
- PRTWA810GNRL
- PRTWA810GNRS
- PRTWA810GNRXXL
- PRTWA810GNRM
- Model:
- A810
- A810
- A810
- A810
- A810
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Textured
- Textured
- Textured
- Textured
- Textured
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X)
- EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X)
- EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X)
- EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X)
- EN ISO 374-1:2016 Type A (AJKLMNOT), EN ISO 374-5 Micro Organisms, EN 388:2016 (3101X)