ContactUsLinkComponent
Wrap around glasses, PW32
Wrap around glasses, PW32
Catalog # PRTWPW32CLR
Supplier:  Portwest
undefined
Wrap around glasses, PW32
Catalog # PRTWPW32CLR
Supplier:  Portwest
Specifications
  • Type:
    UV protection, anti scratch
  • Colour:
    Black
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Wrap-around design glasses with one piece lens.
  • 99% UV protection
  • Anti-scratch coating for added durability
  • Flexible arms for ultra comfort
  • Ultra lightweight for long uses
  • Individually packed for vending machines
  • Retail bag which aids presentation for retail sales
Certifications: AS/NZS 1337.1; EN 170 (2C-1.2 (clear)); ANSI/ISEA Z87 + (U6 (Clear))
Delivery: Cord included.