ContactUsLinkComponent
Face shield, WP98 series
Face shield, WP98 series
Catalog # MMMAWP98
Supplier:  3M
undefined
Face shield, WP98 series
Catalog # MMMAWP98
Supplier:  3M
Specifications
  • Description:
    WP98 series face shield
  • Pk:
    1
Specifications
About this item
The WP98 Series Face Shield is a high impact resistant, clear acetate faceshield designed to fit 3M hard hats. It can be used with the H8 Headgear easy faceshield replacement system.
  • Provides protection against impacts, liquids, and chemical splashes
  • Clear acetate faceshield
  • High impact resistance
  • Can be used with the H8 Headgear easy faceshield replacement system
  • The shield goes inside the attachment for extra support
The WP98 Series clear acetate faceshield protects against high speed particles with low energy impact (0,86 g ball at 45 m/s), marking F. WP98 is certified for use with SecureFit helmets using holder U5B and is certified for use with G3000 and G3501 using holder U3-E. It is also certified for use with H8 Headgear.