Tetrahydrofuran ≥99.8% stabilised, AR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Boiling Pt:65 °C
Melting Pt:–108.3 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm
:GHS02
:GHS07
:GHS08
Specification Test Results
- Assay (GC):≥99,8
- Water (KF):≤0,03%
- Titrable Acid (meq/g):≤0,0005
- Residue after Evaporation:≤0,03%
- Stabiliser (BHT) :~250 ppm